Learn More
World Precision Instrument Wire Mesh Sterilization Baskets, Drop Handles-WP-4404
SDP
Catalog No. 502090247
Encompass
Stainless steel, electropolished Wire Mesh Sterilization Baskets with drop handles for instrument cleaning, sterilization and storage. The baskets meet all requirements of automatic washing and sterilization for all types of instruments.
Catalog No. 50-209-0247 Supplier World Precision Instrument Supplier No. WP4404
Stainless steel, electropolished Wire Mesh Sterilization Baskets with drop handles for instrument cleaning, sterilization and storage. The baskets meet all requirements of automatic washing and sterilization for all types of instruments.