Learn More
World Precision Instrument Wire Mesh Sterilization Baskets, Drop Handles-WP-4403
SDP
Catalog No. 502090244
Encompass
Stainless steel, electropolished Wire Mesh Sterilization Baskets with drop handles for instrument cleaning, sterilization and storage. The baskets meet all requirements of automatic washing and sterilization for all types of instruments.
Catalog No. 50-209-0244 Supplier World Precision Instrument Supplier No. WP4403
Stainless steel, electropolished Wire Mesh Sterilization Baskets with drop handles for instrument cleaning, sterilization and storage. The baskets meet all requirements of automatic washing and sterilization for all types of instruments.