Learn More
World Precision Instrument Iris Scissors, 90mm long, Sharp tips, curved, 5mm, Titanium
SDP
Catalog No. 502090265
Encompass
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Catalog No. 50-209-0265 Supplier World Precision Instrument Supplier No. WP4410
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.