Learn More
World Precision Instrument Wire Mesh Sterilization Baskets, Drop Handles-WP-4401
SDP
Catalog No. 502090238
Encompass
Stainless steel, electropolished Wire Mesh Sterilization Baskets with drop handles for instrument cleaning, sterilization and storage. The baskets meet all requirements of automatic washing and sterilization for all types of instruments.
This item is not returnable. View return policy
Catalog No. 50-209-0238 Supplier World Precision Instrument Supplier No. WP4401
Stainless steel, electropolished Wire Mesh Sterilization Baskets with drop handles for instrument cleaning, sterilization and storage. The baskets meet all requirements of automatic washing and sterilization for all types of instruments.