Learn More
Cole-Parmer Davis Instruments 7627 Solar-powered Repeater
Supplier:  Cole-Parmer 8640343
Encompass_Preferred
Wireless Repeater receives data from any compatible transmitter and re-transmits it to any compatible Vantage Pro2 or Vantage Vue console/+receiver. The repeater extends the transmission range and improves reception between the transmitter and the receiver.
Catalog No. 50-628-454