Learn More
Med Vet International Student Microscope w/ LED & mechanical stage
Catalog No. 503047223
Encompass_Preferred
Description(Model MSK-01L) is the new economy model combining the quality of the My First Lab with the upgrades of the Premiere MS series for greater options. Equipped with the mechanical stage which allows smooth and easy adjustment of slides lieft/right or backward/foward during observation as well as both coarse and fine focusing controls, popular features for educational use. Quality metal construction and a 5 year warranty makes this reliable microscope a great value.10X Eyepiece with scale pointer3 Objectives 4X, 10X, 40XTotal magnification: 40X to 100XMechanical stageCoaxial coarse & fine focusingDisc diaphragmLED LightingSpare bulbConforms to ASTM F963
Catalog No. 50-304-7223 Supplier Med Vet International Supplier No. MSK01L
Description(Model MSK-01L) is the new economy model combining the quality of the My First Lab with the upgrades of the Premiere MS series for greater options. Equipped with the mechanical stage which allows smooth and easy adjustment of slides lieft/right or backward/foward during observation as well as both coarse and fine focusing controls, popular features for educational use. Quality metal construction and a 5 year warranty makes this reliable microscope a great value.10X Eyepiece with scale pointer3 Objectives 4X, 10X, 40XTotal magnification: 40X to 100XMechanical stageCoaxial coarse & fine focusingDisc diaphragmLED LightingSpare bulbConforms to ASTM F963