Learn More
AB Sciex LLC STEADSPRAY ELECTRODE 2PK
Catalog No. NC2207779
Encompass
SteadySpray Electrode High Micro 50-200L/min2-pack of electrodes for use with the OptiFlow Turbo V source. To be used with flow rates 50-200 uL/min. Stainless steel, tan sleeve.
This item is not returnable. View return policy
Catalog No. NC2207779 Supplier AB Sciex LLC Supplier No. 5061576
SteadySpray Electrode High Micro 50-200L/min2-pack of electrodes for use with the OptiFlow Turbo V source. To be used with flow rates 50-200 uL/min. Stainless steel, tan sleeve.