Learn More
Biotium STEADY MEMBRANE STAIN KIT,500
Catalog No. NC1866302
Encompass_Preferred
CellBrite™ Steady Membrane Staining Kits •Full size kit (500 labelings): •CellBrite™ Steady Dye, 1000X in DMSO, 100 uL.•CellBrite™ Steady Enhancer, 1000X in DMSO, 100 uL.CellBrite™ Steady Membrane Staining Kits allow fluorescence imaging of cell surface for up to several days in culture. The CellBrite™ Steady Dyes are unique fluorescent membrane probes that distribute between the cell surface and intracellular compartments, so cells retain cell surface staining over time. With the use of CellBrite™ Steady Enhancer, intracellular staining can be reduced or eliminated for imaging of cell outlines or boundaries.
Catalog No. NC1866302 Supplier Biotium Supplier No. 30106
CellBrite™ Steady Membrane Staining Kits •Full size kit (500 labelings): •CellBrite™ Steady Dye, 1000X in DMSO, 100 uL.•CellBrite™ Steady Enhancer, 1000X in DMSO, 100 uL.CellBrite™ Steady Membrane Staining Kits allow fluorescence imaging of cell surface for up to several days in culture. The CellBrite™ Steady Dyes are unique fluorescent membrane probes that distribute between the cell surface and intracellular compartments, so cells retain cell surface staining over time. With the use of CellBrite™ Steady Enhancer, intracellular staining can be reduced or eliminated for imaging of cell outlines or boundaries.