Learn More
Precisionary Instruments Inc STEADYHOLD TUBE STABILIZER STS
SDP
Catalog No. NC3589762
Encompass_Preferred
Compresstome® SteadyHold Tube Stabilizer (STS) kitThe SteadyHold Tube Stabilizer (STS) is an innovative tool designed to enhance precision and stability when embedding tissue samples in Compresstome vibratome specimen tubes. This stabilizer securely holds specimen tubes in place, allowing for seamless embedding with low-melting point agarose while maintaining sample integrity.
Catalog No. NC3589762 Supplier Precisionary Instruments Inc Supplier No. VFSTEADYHOLDTUBESTABIL
Compresstome® SteadyHold Tube Stabilizer (STS) kitThe SteadyHold Tube Stabilizer (STS) is an innovative tool designed to enhance precision and stability when embedding tissue samples in Compresstome vibratome specimen tubes. This stabilizer securely holds specimen tubes in place, allowing for seamless embedding with low-melting point agarose while maintaining sample integrity.