Learn More
AB Sciex LLC SteadySpray Electrode Low Micro 1-10 uL/min, 5/pk
Catalog No. NC1715021
Encompass
5-pack of electrodes for use with the OptiFlow Turbo V source. To be used with flow rates 1-10 uL/min. Fused silica, red sleeve.
Catalog No. NC1715021 Supplier AB Sciex LLC Supplier No. 5061574
5-pack of electrodes for use with the OptiFlow Turbo V source. To be used with flow rates 1-10 uL/min. Fused silica, red sleeve.