Learn More
AB Sciex LLC STEADYSPRAY MICRO ELECTRODE,
Catalog No. NC1777197
Encompass
SteadySpray Micro Electrode, 10-50uL/min, Beige PEEKsil
Catalog No. NC1777197 Supplier AB Sciex LLC Supplier No. 5061575
SteadySpray Micro Electrode, 10-50uL/min, Beige PEEKsil