Learn More
MSC Uvex Safety Goggles, Anti-Fog Polycarbonate, Indirect Vent, Gray, 1 ea
Catalog No. 502370348
Encompass
These safety goggles provide essential eye protection with a focus on clear vision and comfort. Designed for use in environments requiring protection from dust and splash, they feature advanced anti-fog lenses and an indirect vent system to maintain clarity in challenging conditions.
  • Anti-fog coatings improve visibility under hot and humid conditions
  • Clear polycarbonate lenses offer impact resistance
  • Indirect vent design for dust and splash protection
  • Gray elastomer frame ensures a comfortable, universal fit
  • Provides UV protection
  • Meets ANSI Z87.1+ standards
  • Includes a neoprene headband for secure wear
Catalog No. 50-237-0348 Supplier MSC Supplier No. 53160115
These safety goggles provide essential eye protection with a focus on clear vision and comfort. Designed for use in environments requiring protection from dust and splash, they feature advanced anti-fog lenses and an indirect vent system to maintain clarity in challenging conditions.
  • Anti-fog coatings improve visibility under hot and humid conditions
  • Clear polycarbonate lenses offer impact resistance
  • Indirect vent design for dust and splash protection
  • Gray elastomer frame ensures a comfortable, universal fit
  • Provides UV protection
  • Meets ANSI Z87.1+ standards
  • Includes a neoprene headband for secure wear