Learn More
Electron Microscopy Sciences Steam Autoclave Printed Tape (B), 1" wide, 3" core 60 YD
SDP
Catalog No. 5025392
Encompass_Preferred
Indicate the status of all laboratory items to be sterilized with these tape labels. The word "STERILIZED" appears after processing at 250 degree F under normal sterilization conditions.
Width: 1"
Core: 3" core
Catalog No. 50-253-92 Supplier Electron Microscopy Sciences Supplier No. 7701710
Indicate the status of all laboratory items to be sterilized with these tape labels. The word "STERILIZED" appears after processing at 250 degree F under normal sterilization conditions.
Width: 1"
Core: 3" core