Learn More
Ace Glass, Inc. Steam distillation condenser only
SDP
Catalog No. 50973256
Encompass
60 x 500mm steam distillation condenser with 24/40 joints and a solvent capacity of 15mL.
Catalog No. 50-973-256 Supplier Ace Glass, Inc. Supplier No. 655540
60 x 500mm steam distillation condenser with 24/40 joints and a solvent capacity of 15mL.