Learn More
SPS Medical Indicator Label w/ operator and date 0.75" x 1.25", 250/roll
Catalog No. NC0914758
Encompass
Crosstex Steam Chemical Process Indicators (CPIs) are manufactured to monitor whether Steam exposure conditions were met at the point of application using pressure sensitive adhesive. STL-250 are intended for use with individual units (e.g. packs, containers) to indicate that the unit has been directly exposed to a Steam process and to distinguish between processed and unprocessed units.
Catalog No. NC0914758 Supplier SPS Medical Supplier No. STL250
Crosstex Steam Chemical Process Indicators (CPIs) are manufactured to monitor whether Steam exposure conditions were met at the point of application using pressure sensitive adhesive. STL-250 are intended for use with individual units (e.g. packs, containers) to indicate that the unit has been directly exposed to a Steam process and to distinguish between processed and unprocessed units.