Learn More
Amsco Scientific Steris Steam Integrating Indicator 5CM, 500/pk
Catalog No. NC1896692
Encompass
The VERIFY STEAM Integrating Indicator 5CM is designed to verify following a sterilization process that critical steam sterilization parameters were met. The 2-inch long Type 5 Integrating Indicator fits where other longer chemical indicators do not, all while meeting the stringent performance requirements of ANSI/AAMI/ISO chemical indicator standard 11140-1.
This item is not returnable. View return policy
Catalog No. NC1896692 Supplier Amsco Scientific Steris Supplier No. PCC067
The VERIFY STEAM Integrating Indicator 5CM is designed to verify following a sterilization process that critical steam sterilization parameters were met. The 2-inch long Type 5 Integrating Indicator fits where other longer chemical indicators do not, all while meeting the stringent performance requirements of ANSI/AAMI/ISO chemical indicator standard 11140-1.