Learn More
Andwin Scientific 3M™ Attest™ Steam Chemical Integrator
SDP
Catalog No. NC9265285
Encompass_Preferred
Moving front ink technology with vibrant "Accept" (Green) and "Reject" (Red) zones for easy interpretation within seconds. 3M™ Attest™ White/Red colored back: Makes the Chemical Integrator easy to find if/when it flips over within a surgical tray (Category 5) Integrating Indicators. The integrator is designed to correlate with the performance of a BI at 3 time/temperature relationships*
Catalog No. NC9265285 Supplier Andwin Scientific Supplier No. 4135
Moving front ink technology with vibrant "Accept" (Green) and "Reject" (Red) zones for easy interpretation within seconds. 3M™ Attest™ White/Red colored back: Makes the Chemical Integrator easy to find if/when it flips over within a surgical tray (Category 5) Integrating Indicators. The integrator is designed to correlate with the performance of a BI at 3 time/temperature relationships*