Learn More
McKesson General Medical Comply SteriGage Steam Chem Integrator b
Catalog No. NC1141832
Encompass
INTEGRATOR, STEAM, MOVING FRONT, 2"X3/4" Moving front ink technology provides easy to read 'Accept or Reject' result. Can be used in all 250-275°F steam sterilization cycles. Response correlates with a Biological Indicator at 3 time/temperature relationships under ideal steam sterilization conditions. 500/bag. 2 bags for 1000.
Catalog No. NC1141832 Supplier McKesson General Medical Supplier No. 134499
Add to Cart
Add to Cart
INTEGRATOR, STEAM, MOVING FRONT, 2"X3/4" Moving front ink technology provides easy to read 'Accept or Reject' result. Can be used in all 250-275°F steam sterilization cycles. Response correlates with a Biological Indicator at 3 time/temperature relationships under ideal steam sterilization conditions. 500/bag. 2 bags for 1000.