Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lux Blox™ STEAM Inventor with Accessories
SCI_ED

Catalog No. S81700
Change view
Click to view available options
Grade:
Pre-K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S81700 Pre-K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Set Includes Small Parts Which May Present a Choking Hazard
Catalog No. S81700 Supplier Lux Blox™ Supplier No. LUXSI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Set Includes Small Parts Which May Present a Choking Hazard

A big, beautiful tube of inventing fun! Certified as an authentic STEM experience, kids can enter into the world of LUX construction and learn how to build big working machines, creatures, or their own toys and inventions.

  • Kit includes 223 pieces
  • Contains small parts which may present a choking hazard
  • Packaging: Clear, 14 in. Tube

Specifications

Grade Pre-K to 12
Age 6
Includes Lux Squares (170), Lux Trigons (35), Square Multi-Wheels (4), Trigon Multi-Wheels (4), Lux Axles (10), and 7 in. Rubber Bands (2)
Quantity 1 Kit
Product Type STEAM Inventor
Description Contains Enough Pieces to Construct One Item at a Time
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.