Learn More
Biocare Medical, LLC Steam Monitor Strips - 250 Strips
SDP
Catalog No. 5082357
Encompass_Preferred
Heat-induced epitope retrieval methods (HIER) are used to pretreat tissues for optimum antibody-antigen reaction. Many different kinds of heat retrieval methods are used. These include the following: microwave, waterbath, steamers and pressure cooker methods. These methods were developed to “Standardize” immunohistochemistry for formalin-fixed, paraffin-embedded tissues. Heat retrieval methods’ lack of “standardization” and quality control (QC) procedures is a problem. BIOCARE QC Steam Monitor Strips are for antigen retrieval in a pressure steam chamber (Decloaking Chamber). The Steam Monitor Strip gives a visual QC to make sure that the pressure cooker has reached the optimum temperature and pressure.
Catalog No. 50-823-57 Supplier Biocare Medical, LLC Supplier No. 613D
Heat-induced epitope retrieval methods (HIER) are used to pretreat tissues for optimum antibody-antigen reaction. Many different kinds of heat retrieval methods are used. These include the following: microwave, waterbath, steamers and pressure cooker methods. These methods were developed to “Standardize” immunohistochemistry for formalin-fixed, paraffin-embedded tissues. Heat retrieval methods’ lack of “standardization” and quality control (QC) procedures is a problem. BIOCARE QC Steam Monitor Strips are for antigen retrieval in a pressure steam chamber (Decloaking Chamber). The Steam Monitor Strip gives a visual QC to make sure that the pressure cooker has reached the optimum temperature and pressure.