Learn More
Grainger Steam Pan Liner 18 in W x 14 in L PK250
Catalog No. 502750840
Encompass_Preferred
The Daymark Disposable Steam Pan Liner is a disposable liner designed to fit standard 12" x 20" x 2.5" steam table pans. It is made from polyethylene film and can withstand temperatures from -40°F to 200°F. The liner measures 12.25" x 20.25" x 2.5" and has a capacity of 4 quarts. It features a food-grade, leak-resistant seal and is intended for single use to simplify cleanup. This USA-made liner comes in packs of 250. It provides a hygienic barrier between food and pans.
Catalog No. 50-275-0840 Supplier Grainger Supplier No. 14H146
The Daymark Disposable Steam Pan Liner is a disposable liner designed to fit standard 12" x 20" x 2.5" steam table pans. It is made from polyethylene film and can withstand temperatures from -40°F to 200°F. The liner measures 12.25" x 20.25" x 2.5" and has a capacity of 4 quarts. It features a food-grade, leak-resistant seal and is intended for single use to simplify cleanup. This USA-made liner comes in packs of 250. It provides a hygienic barrier between food and pans.