Learn More
SPS Medical STEAM + PLUS STERIL INTEGRATER
Catalog No. NC9860745
Encompass
STEAMPlus Integrator, 100/pk , 10/case, Scientifically engineered to integrate all 3 critical parameters of sterilization, For use in ALL Steam (Gravity, vacuum and flash) sterilization processes
Catalog No. NC9860745 Supplier SPS Medical Supplier No. SSI100
STEAMPlus Integrator, 100/pk , 10/case, Scientifically engineered to integrate all 3 critical parameters of sterilization, For use in ALL Steam (Gravity, vacuum and flash) sterilization processes