Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lux Blox™ STEAM Primer with Accessories
SCI_ED

Catalog No. S81699
Change view
Click to view available options
Grade:
Pre-K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S81699 Pre-K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Set Includes Small Parts Which May Present a Choking Hazard
Catalog No. S81699 Supplier Lux Blox™ Supplier No. LUXSP
Add to Cart
Add to Cart
Set Includes Small Parts Which May Present a Choking Hazard

A revolution in smart construction fun! Keep young imaginations entertained for hours and immerse them into the STEAM arts of science, technology, engineering, art, and math!

  • Kit includes 111 pieces
  • Contains small parts which may present a choking hazard

Specifications

Color Assorted
Grade Pre-K to 12
Age 6
Includes Lux Squares (75), White Trigons (20), Trigon Wheels (5), Square Wheels (4), Lux Axels (5), and 7 in. Rubber Bands (2)
Quantity 1 Kit
Product Type STEAM Primer
Description Contains Enough Pieces to Construct One Item at a Time
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.