Learn More
Carltex Inc STEAM QUALITY TEST KIT
SDP
Catalog No. NC1792715
Encompass
  • Integrated SQ1E NCG condenser unit
  • Air-cooled condenser type
  • Measuring parameters: Dryness value, superheat, non-condensable gases (NCG)
  • Easy to use with detailed manual
  • Robust, light, and compact design
  • Wheeled carrying case for portability
  • Folding attachable table for convenience in fieldwork
  • Flow pause button to prevent sample loss during re-zeroing
  • Burette features: Integral, push-button zeroing, back-lighting
  • Compliant with EN 285 standard
  • Repeatable and assured test results
  • Target industries: Pharma, biotech, healthcare
Catalog No. NC1792715 Supplier Carltex Inc Supplier No. SQ1E
Add to Cart
Add to Cart
Request a Quote
  • Integrated SQ1E NCG condenser unit
  • Air-cooled condenser type
  • Measuring parameters: Dryness value, superheat, non-condensable gases (NCG)
  • Easy to use with detailed manual
  • Robust, light, and compact design
  • Wheeled carrying case for portability
  • Folding attachable table for convenience in fieldwork
  • Flow pause button to prevent sample loss during re-zeroing
  • Burette features: Integral, push-button zeroing, back-lighting
  • Compliant with EN 285 standard
  • Repeatable and assured test results
  • Target industries: Pharma, biotech, healthcare