Learn More
CROWN FOOD SERVICE EQUIPMENT Market Forge Sterilmatic Sterilizer Autoclave, Digital,W/Temp Control 230-250F, 208/240V 60Hz
Catalog No. NC2180821
Encompass_Preferred
Sterilize Glass Beakers, Pipettes, Test Tubes And Flasks. Metal And Plastic Instruments, Syringes, Needles, Brushes, Rubber Gloves, Catheters, Rubber Tubing, Dressings, Linens, Petri Dishes And More. Standard Features: Lcd Display Touch ScreenTemperature In °F Or °C And Pressure In Psi Or KpaSterilizing Temperatures From 225-250°F (107-121°C)Choose From Fast Or Slow ExhaustBuilt-In Usb Port And Rs232 Serial Port For PrinterProgramable Controller To Store Multiple Temp/Time/Vent SettingsMaintains Set Temperature With Built-In MicroprocessorElectric Steam TrapRemovable Pan SupportsManual Fill
Catalog No. NC2180821 Supplier CROWN FOOD SERVICE EQUIPMENT Supplier No. STMED
Add to Cart
Add to Cart
Request a Quote
Sterilize Glass Beakers, Pipettes, Test Tubes And Flasks. Metal And Plastic Instruments, Syringes, Needles, Brushes, Rubber Gloves, Catheters, Rubber Tubing, Dressings, Linens, Petri Dishes And More. Standard Features: Lcd Display Touch ScreenTemperature In °F Or °C And Pressure In Psi Or KpaSterilizing Temperatures From 225-250°F (107-121°C)Choose From Fast Or Slow ExhaustBuilt-In Usb Port And Rs232 Serial Port For PrinterProgramable Controller To Store Multiple Temp/Time/Vent SettingsMaintains Set Temperature With Built-In MicroprocessorElectric Steam TrapRemovable Pan SupportsManual Fill