Learn More
COLDER PRODUCTS COMPANY CPC Autoclavable Sanitary SIP Connectors Includes 1/2 hose barb x 3/4 x 1-1/2 in sanitary SIP connector Free floating Seal Silicone platinum-cured USP Class VI ADCF SIP process
Catalog No. 503131468
Encompass_Preferred
CPC Autoclavable Sanitary SIP Connectors Includes 1/2 hose barb x 3/4 x 1-1/2 in sanitary SIP connector Free floating Seal Silicone platinum-cured USP Class VI ADCF SIP process
Catalog No. 50-313-1468 Supplier COLDER PRODUCTS COMPANY Supplier No. STC1700700
CPC Autoclavable Sanitary SIP Connectors Includes 1/2 hose barb x 3/4 x 1-1/2 in sanitary SIP connector Free floating Seal Silicone platinum-cured USP Class VI ADCF SIP process