Learn More
MSC PortionPac Carpet Extraction Detergent, CRI Certified, Green Seal Certified
Catalog No. 502838555
Encompass
PortionPac SteamPac Carpet Extraction Detergent is a powerful yet gentle formulation suitable for extraction or bonnet carpet cleaning. It combines pre-spotting agents, odor counteractants, and de-foaming agents into one high-performance extraction detergent. The formulation is biodegradable and VOC-free, making it an effective and environmentally conscious choice for carpet maintenance.
  • Powerful yet gentle formulation for extraction or bonnet carpet cleaning.
  • Combines pre-spotting agents, odor counteractants, and de-foaming agents.
  • Biodegradable and VOC-free.
  • Carpet and Rug Institute Seal of Approval certified.
  • Meets Green Seal Standard GS-37.
  • No hazards as defined by OSHA Hazard Communication Standard.
Catalog No. 50-283-8555 Supplier MSC Supplier No. 38691085
PortionPac SteamPac Carpet Extraction Detergent is a powerful yet gentle formulation suitable for extraction or bonnet carpet cleaning. It combines pre-spotting agents, odor counteractants, and de-foaming agents into one high-performance extraction detergent. The formulation is biodegradable and VOC-free, making it an effective and environmentally conscious choice for carpet maintenance.
  • Powerful yet gentle formulation for extraction or bonnet carpet cleaning.
  • Combines pre-spotting agents, odor counteractants, and de-foaming agents.
  • Biodegradable and VOC-free.
  • Carpet and Rug Institute Seal of Approval certified.
  • Meets Green Seal Standard GS-37.
  • No hazards as defined by OSHA Hazard Communication Standard.