Learn More
Aviva Systems Biology Corp STEAP1 Antibody - N-terminal region (ARP36697_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36697_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STEAP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human STEAP1. Alternate names: MGC19484, PRSS24, STEAPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-512