Learn More
Enzo Life Sciences STEAP1 monoclonal antibody (J2D2) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS7410100
Encompass_Preferred
Clone: J2D2. Isotype: IgG2bκ. Immunogen: Recombinant human STEAP1 (aa 1-70) purified from E. coli. Source: Purified from ascites. Species reactivity: Human. Applications: ELISA, ICC, IF, IHC, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 0.02% sodium azide and 10% glycerol.
Catalog No. 50-201-3282