Learn More
OriGene STEAP1B CytoSection, 25 Slides
SDP
Catalog No. NC3734725
Encompass_Preferred
Transient overexpression of STEAP1B, transcript variant 1, in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack
Catalog No. NC3734725 Supplier OriGene Supplier No. TS428274P5
Transient overexpression of STEAP1B, transcript variant 1, in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack