Learn More
Aviva Systems Biology Corp STEAP3 Antibody - C-terminal region (ARP43515_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43515_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STEAP3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human STEAP3. Alternate names: STMP3, TSAP6, dudlin-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53366