Learn More
Aviva Systems Biology Corp STEAP4 Antibody - C-terminal region (ARP49842_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49842_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STEAP4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human STEAP4. Alternate names: DKFZp666D049, FLJ23153, STAMP2, TIARP, TNFAIP9ApplicationWB. Species reactivity: Cow, Human, Rabbit
Catalog No. 50-014-60198