Learn More
OriGene STEAP4 CytoSection 25 slides
SDP
Catalog No. NC3734421
Encompass_Preferred
Transient overexpression of STEAP4, transcript variant 1, in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack
Catalog No. NC3734421 Supplier OriGene Supplier No. TS416917P5
Transient overexpression of STEAP4, transcript variant 1, in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack