Learn More
Medchemexpress LLC Stearamide | 124-26-5 | MFCD00008038 | 97.0% | 283.49 | C18H37NO | 250 MG
SDP
Catalog No. 5000283481
Encompass_Preferred
Stearamide is a primary fatty acid amide for research use. It displays cytotoxic and ichthytoxic activity and has been reported to form an adsorption layer on carbon steel, providing corrosion inhibition; cellular studies indicate it can stimulate protein synthesis via the PI3K/Akt/mTOR pathway.
  • Primary fatty acid amide with documented cytotoxic and ichthytoxic activity.
  • High chemical purity (97.0%).
  • White to off-white solid physical form.
  • Soluble in ethanol: 12.5 mg/mL (requires sonication).
  • Stable when stored as powder: -20°C (up to 3 years) or 4°C (up to 2 years).
  • Suitable for corrosion inhibition and cellular signaling research applications.
Catalog No. 50-002-83481 Supplier Medchemexpress LLC Supplier No. HYW130610250MG
Stearamide is a primary fatty acid amide for research use. It displays cytotoxic and ichthytoxic activity and has been reported to form an adsorption layer on carbon steel, providing corrosion inhibition; cellular studies indicate it can stimulate protein synthesis via the PI3K/Akt/mTOR pathway.
  • Primary fatty acid amide with documented cytotoxic and ichthytoxic activity.
  • High chemical purity (97.0%).
  • White to off-white solid physical form.
  • Soluble in ethanol: 12.5 mg/mL (requires sonication).
  • Stable when stored as powder: -20°C (up to 3 years) or 4°C (up to 2 years).
  • Suitable for corrosion inhibition and cellular signaling research applications.