Learn More
Medchemexpress LLC Stearamide | 124-26-5 | MFCD00008038 | 97.0% | 283.49 g/mol | C18H37NO | 25 G
SDP
Catalog No. 5000283312
Encompass_Preferred
Stearamide is a primary fatty acid amide used as a research reagent and corrosion inhibitor. It has reported cytotoxic and ichthyotoxic activity and is supplied for laboratory research use only. Purity is typically listed as 97.0%.
  • Primary fatty acid amide chemical.
  • Reported cytotoxic and ichthyotoxic activity.
  • Commonly used as a corrosion inhibitor in materials research.
  • Supplied for laboratory research use only (not for human use).
  • High purity suitable for analytical and experimental applications.
Catalog No. 50-002-83312 Supplier Medchemexpress LLC Supplier No. HYW13061025G
Stearamide is a primary fatty acid amide used as a research reagent and corrosion inhibitor. It has reported cytotoxic and ichthyotoxic activity and is supplied for laboratory research use only. Purity is typically listed as 97.0%.
  • Primary fatty acid amide chemical.
  • Reported cytotoxic and ichthyotoxic activity.
  • Commonly used as a corrosion inhibitor in materials research.
  • Supplied for laboratory research use only (not for human use).
  • High purity suitable for analytical and experimental applications.