Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Stearamide | 124-26-5 | MFCD00008038 | 97.0% | 283.49 g/mol | C18H37NO | 500 MG
SDP

Catalog No. 5000283547
Encompass_Preferred

Stearamide is a primary fatty acid amide (octadecanamide) supplied as a research-grade reagent. It has reported cytotoxic and ichthyotoxic activity and is used in chemical research, materials studies, and analytical applications. Available in small laboratory pack sizes for experimental work.

  • Primary fatty acid amide (octadecanamide).
  • CAS number 124-26-5.
  • Molecular formula C18H37NO; molecular weight 283.49 g/mol.
  • Typical purity around 97.0%.
  • Available in laboratory pack sizes including 500 mg.
  • Suitable for research and material testing applications.

Catalog No. 50-002-83547 Supplier Medchemexpress LLC Supplier No. HYW130610500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stearamide is a primary fatty acid amide (octadecanamide) supplied as a research-grade reagent. It has reported cytotoxic and ichthyotoxic activity and is used in chemical research, materials studies, and analytical applications. Available in small laboratory pack sizes for experimental work.

  • Primary fatty acid amide (octadecanamide).
  • CAS number 124-26-5.
  • Molecular formula C18H37NO; molecular weight 283.49 g/mol.
  • Typical purity around 97.0%.
  • Available in laboratory pack sizes including 500 mg.
  • Suitable for research and material testing applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.