Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Stearamide | 124-26-5 | MFCD00008038 | 97.0% | 283.5 g/mol | C18H37NO | 250 MG
SDP

Catalog No. 5000283284
Encompass_Preferred

Stearamide is a primary fatty acid amide (CAS 124-26-5) supplied as a research-grade analytical standard. It is provided as a white to off-white solid and is used in analytical chemistry, corrosion-inhibition studies, and biochemical assays where activity on PI3K/Akt/mTOR signaling has been reported. For research use only.

  • Primary fatty acid amide suitable as an analytical standard.
  • CAS number 124-26-5 for unambiguous identification.
  • Purity approximately 97.0%.
  • White to off-white solid form, easy to handle and weigh.
  • Forms an adsorption layer on carbon steel; applicable to corrosion studies.
  • Reported biological activity in PI3K/Akt/mTOR signaling for cell assays.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-83284 Supplier Medchemexpress LLC Supplier No. HYW130610R250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stearamide is a primary fatty acid amide (CAS 124-26-5) supplied as a research-grade analytical standard. It is provided as a white to off-white solid and is used in analytical chemistry, corrosion-inhibition studies, and biochemical assays where activity on PI3K/Akt/mTOR signaling has been reported. For research use only.

  • Primary fatty acid amide suitable as an analytical standard.
  • CAS number 124-26-5 for unambiguous identification.
  • Purity approximately 97.0%.
  • White to off-white solid form, easy to handle and weigh.
  • Forms an adsorption layer on carbon steel; applicable to corrosion studies.
  • Reported biological activity in PI3K/Akt/mTOR signaling for cell assays.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.