Learn More
Medchemexpress LLC Stearate sodium 100g
SDP
Catalog No. 5000205607
Encompass_Preferred
Stearate sodiumIt is an organic compound belonging to fatty acids It is the sodium salt of stearic acid a saturated fatty acid commonly found in various animal and vegetable fats Stearate sodiumCommonly used as surfactants emulsifiers and detergents it is widely used in the cosmetics personal care and detergent industries It s found in a variety of products including soaps shampoos toothpaste and laundry detergent Also it has applications in the food industry where it is used as a food additive to improve texture and shelf life
Catalog No. 50-002-05607 Supplier Medchemexpress LLC Supplier No. HYW099570100G
Stearate sodiumIt is an organic compound belonging to fatty acids It is the sodium salt of stearic acid a saturated fatty acid commonly found in various animal and vegetable fats Stearate sodiumCommonly used as surfactants emulsifiers and detergents it is widely used in the cosmetics personal care and detergent industries It s found in a variety of products including soaps shampoos toothpaste and laundry detergent Also it has applications in the food industry where it is used as a food additive to improve texture and shelf life