Learn More
Cayman Chemical Stearic Acid glycidyl ester, Purity: ≥95%, CAS: 7460-84-6, 100 mg
SDP
Catalog No. 502909322
Encompass_Preferred
An esterified form of stearic acid; induces the formation of subcutaneous sarcomas and pulmonary tumors in mice from 0.005-10 mg/animal; induces the formation of breast carcinomas in mice at 0.1 mg/animal; serum levels are decreased in patients with prostate cancer compared to patients with benign prostatic hyperplasia and are negatively correlated with total cholesterol, LDL, and ApoB levels
Catalog No. 50-290-9322 Supplier Cayman Chemical Supplier No. 36838100 MG
An esterified form of stearic acid; induces the formation of subcutaneous sarcomas and pulmonary tumors in mice from 0.005-10 mg/animal; induces the formation of breast carcinomas in mice at 0.1 mg/animal; serum levels are decreased in patients with prostate cancer compared to patients with benign prostatic hyperplasia and are negatively correlated with total cholesterol, LDL, and ApoB levels