Learn More
Selleck Chemical LLC Stearic acid
SDP
Catalog No. 5000003403
Encompass_Preferred
Stearic acid (Octadecanoic acid Isostearic acid Cetylacetic acid) is a natural saturated fatty acid found in animal and vegetable fats It could be used as a food additive and used in soaps cosmetics and detergents
Catalog No. 50-000-03403 Supplier Selleck Chemical LLC Supplier No. S57331G
Stearic acid (Octadecanoic acid Isostearic acid Cetylacetic acid) is a natural saturated fatty acid found in animal and vegetable fats It could be used as a food additive and used in soaps cosmetics and detergents