Learn More
Cayman Chemical StearIc AcId-13C 100mg
SDP
Catalog No. 502938984
Encompass_Preferred
An internal standard for the quantification of stearic acid by GC- or LC-MS
Catalog No. 50-293-8984 Supplier Cayman Chemical Supplier No. 28917100 MG
An internal standard for the quantification of stearic acid by GC- or LC-MS