Learn More
Cayman Chemical Stearic Acid-13C, Purity: ≥95%, CAS: 85541-42-0, 250 mg
SDP
Catalog No. 502939994
Encompass_Preferred
An internal standard for the quantification of stearic acid by GC- or LC-MS
Catalog No. 50-293-9994 Supplier Cayman Chemical Supplier No. 28917250 MG
An internal standard for the quantification of stearic acid by GC- or LC-MS