Learn More
Medchemexpress LLC Stearic acid | 57-11-4 | 99.9% | 10 G
SDP
Catalog No. 5000410185
Encompass_Preferred
Stearic acid is a long-chain dietary saturated fatty acid that significantly reduces visceral fat by inducing apoptosis of preadipocytes. It is utilized in the study of cardiovascular and metabolic diseases.
  • Target: Endogenous metabolite, Apoptosis
  • Research areas: Cancer, Cardiovascular disease, Metabolic disease
Catalog No. 50-004-10185 Supplier Medchemexpress LLC Supplier No. HYB221910G
Stearic acid is a long-chain dietary saturated fatty acid that significantly reduces visceral fat by inducing apoptosis of preadipocytes. It is utilized in the study of cardiovascular and metabolic diseases.
  • Target: Endogenous metabolite, Apoptosis
  • Research areas: Cancer, Cardiovascular disease, Metabolic disease