Learn More
Medchemexpress LLC Stearic acid | 57-11-4 | MFCD00002752 | 99.9% | 1 ML
SDP
Catalog No. 5000410385
Encompass_Preferred
Stearic acid is a long-chain dietary saturated fatty acid that significantly reduces visceral fat by inducing apoptosis of preadipocytes. This compound is valuable for research into cardiovascular and metabolic diseases.
  • Significantly reduces visceral fat
  • Induces apoptosis of preadipocytes
  • Used in cardiovascular and metabolic disease studies
  • Increases apoptosis of 3T3L1 preadipocytes
  • Decreases expression of cIAP2 and Bcl2
  • Can reduce bone mineral density
  • Can reduce abdominal fat content
  • Can reduce serum glucose concentration
Catalog No. 50-004-10385 Supplier Medchemexpress LLC Supplier No. HYB221910MM/1ML
Stearic acid is a long-chain dietary saturated fatty acid that significantly reduces visceral fat by inducing apoptosis of preadipocytes. This compound is valuable for research into cardiovascular and metabolic diseases.
  • Significantly reduces visceral fat
  • Induces apoptosis of preadipocytes
  • Used in cardiovascular and metabolic disease studies
  • Increases apoptosis of 3T3L1 preadipocytes
  • Decreases expression of cIAP2 and Bcl2
  • Can reduce bone mineral density
  • Can reduce abdominal fat content
  • Can reduce serum glucose concentration