Learn More
Cayman Chemical Stearic Acid-9,10-d2, Purity: ≥99% deuterated forms (d1-d2), CAS: 57396-97-1, 10 mg
SDP
Catalog No. 502940356
Encompass_Preferred
An internal standard for the quantification of stearic acid by GC- or LC-MS
Catalog No. 50-294-0356 Supplier Cayman Chemical Supplier No. 2886810 MG
An internal standard for the quantification of stearic acid by GC- or LC-MS