Learn More
Cayman Chemical StearIc AcId-9 10-d2 1mg
SDP
Catalog No. 502939893
Encompass_Preferred
An internal standard for the quantification of stearic acid by GC- or LC-MS
Catalog No. 50-293-9893 Supplier Cayman Chemical Supplier No. 288681 MG
Add to Cart
Add to Cart
An internal standard for the quantification of stearic acid by GC- or LC-MS