Learn More
Cayman Chemical Stearic Acid-9,10-d2, Purity: ≥99% deuterated forms (d1-d2), CAS: 57396-97-1, 25 mg
SDP
Catalog No. 502941854
Encompass_Preferred
An internal standard for the quantification of stearic acid by GC- or LC-MS
Catalog No. 50-294-1854 Supplier Cayman Chemical Supplier No. 2886825 MG
An internal standard for the quantification of stearic acid by GC- or LC-MS