Learn More
Medchemexpress LLC Stearic acid-d2 | 19905-58-9 | 99.1% | 100 MG
SDP
Catalog No. 5000410187
Encompass_Preferred
Stearic acid-d2 is the deuterium labeled Stearic acid, a long chain dietary saturated fatty acid found in animal and vegetable fats and oils. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer for various applications
  • Serves as an internal standard for quantitative analysis
  • Deuterium substitution can affect pharmacokinetic and metabolic profiles
Catalog No. 50-004-10187 Supplier Medchemexpress LLC Supplier No. HYB2219S1100MG
Stearic acid-d2 is the deuterium labeled Stearic acid, a long chain dietary saturated fatty acid found in animal and vegetable fats and oils. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer for various applications
  • Serves as an internal standard for quantitative analysis
  • Deuterium substitution can affect pharmacokinetic and metabolic profiles