Learn More
Cayman Chemical Stearic Acid-d2, Purity: ≥99% deuterated forms (d1-d2), CAS: 19905-58-9, 250 mg
SDP
Catalog No. 502939646
Encompass_Preferred
An internal standard for the quantification of stearic acid by GC- or LC-MS
Catalog No. 50-293-9646 Supplier Cayman Chemical Supplier No. 28150250 MG
An internal standard for the quantification of stearic acid by GC- or LC-MS