Learn More
Cayman Chemical StearIc AcId-d2 500mg
SDP
Supplier:  Cayman Chemical 28150500 MG
Encompass_Preferred
An internal standard for the quantification of stearic acid by GC- or LC-MS
Catalog No. 50-293-8402